Assay Details
| Gene Symbol: LOC4338556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783806.2 | XP_015639292.1 | 12 - 13 | 1607 | 75 | |
| XM_015783807.2 | XP_015639293.1 | 12 - 13 | 1703 | 75 | ||
| XM_015783808.2 | XP_015639294.1 | 12 - 13 | 1660 | 75 | ||
| GenBank mRNA | AK105546.1 | - | 12 - 13 | 1382 | 75 | |
More Information
Set Membership: |
|

LOC4338556
Genomic Map