Assay Details
| Gene Symbol: LOC4338556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783806.2 | XP_015639292.1 | 4 - 5 | 647 | 82 | |
| XM_015783807.2 | XP_015639293.1 | 4 - 5 | 743 | 82 | ||
| XM_015783808.2 | XP_015639294.1 | 4 - 5 | 700 | 82 | ||
More Information
Set Membership: |
|

LOC4338556
Genomic Map