Assay Details
| Gene Symbol: LOC4338644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782141.2 | XP_015637627.1 | 4 - 5 | 1363 | 85 | |
| GenBank mRNA | AK060054.1 | - | 4 - 5 | 782 | 85 | |
| AK068231.1 | - | 1 - 2 | 227 | 85 | ||
| AK098879.1 | - | 4 - 5 | 783 | 85 | ||
| AK104639.1 | - | 4 - 5 | 782 | 85 | ||
More Information
Set Membership: |
|

LOC4338644
Genomic Map