Assay Details
| Gene Symbol: LOC4338820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782951.2 | XP_015638437.1 | 4 - 5 | 1002 | 119 | |
| GenBank mRNA | AK100958.1 | - | 4 - 5 | 987 | 119 | |
More Information
Set Membership: |
|

LOC4338820
Genomic Map