Assay Details
| Gene Symbol: LOC4338867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782456.2 | XP_015637942.1 | 7 - 8 | 1579 | 62 | |
| GenBank mRNA | AK061594.1 | - | 3 - 4 | 666 | 62 | |
| JN989970.1 | - | 7 - 8 | 1532 | 62 | ||
More Information
Set Membership: |
|

LOC4338867
Genomic Map