Assay Details
| Gene Symbol: LOC4338867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782456.2 | XP_015637942.1 | 8 - 9 | 1772 | 60 | |
| GenBank mRNA | AK061594.1 | - | 4 - 5 | 859 | 60 | |
| JN989970.1 | - | 8 - 9 | 1725 | 60 | ||
More Information
Set Membership: |
|

LOC4338867
Genomic Map