Assay Details
| Gene Symbol: LOC4338867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782456.2 | XP_015637942.1 | 10 - 11 | 2452 | 63 | |
| GenBank mRNA | AK061594.1 | - | 6 - 7 | 1539 | 63 | |
| JN989970.1 | - | 10 - 11 | 2405 | 63 | ||
More Information
Set Membership: |
|

LOC4338867
Genomic Map