Assay Details
| Gene Symbol: LOC4339007 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782700.2 | XP_015638186.1 | 10 - 11 | 1961 | 115 | |
| XM_015782701.2 | XP_015638187.1 | 10 - 11 | 1684 | 115 | ||
| GenBank mRNA | AK073413.1 | - | 10 - 11 | 1691 | 115 | |
| AK104040.1 | - | 8 - 9 | 1170 | 115 | ||
More Information
Set Membership: |
|

LOC4339007
Genomic Map