Assay Details
| Gene Symbol: LOC4339007 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782700.2 | XP_015638186.1 | 4 - 5 | 982 | 98 | |
| XM_015782701.2 | XP_015638187.1 | 4 - 5 | 780 | 98 | ||
| GenBank mRNA | AK073413.1 | - | 4 - 5 | 787 | 98 | |
| AK104040.1 | - | 2 - 3 | 266 | 98 | ||
More Information
Set Membership: |
|

LOC4339007
Genomic Map