Assay Details
| Gene Symbol: LOC4339160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783254.2 | XP_015638740.1 | 3 - 4 | 2365 | 67 | |
| GenBank mRNA | AK070533.1 | - | 3 - 4 | 913 | 67 | |
| BT129156.1 | - | 2 - 3 | 174 | 67 | ||
More Information
Set Membership: |
|

LOC4339160
Genomic Map