Assay Details
| Gene Symbol: LOC4339160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783254.2 | XP_015638740.1 | 6 - 7 | 2914 | 64 | |
| GenBank mRNA | AK070533.1 | - | 6 - 7 | 1462 | 64 | |
More Information
Set Membership: |
|

LOC4339160
Genomic Map