Assay Details
| Gene Symbol: LOC4339189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783826.2 | XP_015639312.1 | 5 - 6 | 675 | 76 | |
| GenBank mRNA | AB288048.1 | - | 5 - 6 | 368 | 76 | |
| AK068320.1 | - | 5 - 6 | 482 | 76 | ||
More Information
Set Membership: |
|

LOC4339189
Genomic Map