Assay Details
| Gene Symbol: LOC4339215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783876.2 | XP_015639362.1 | 25 - 26 | 2112 | 63 | |
| XM_015783878.2 | XP_015639364.1 | 24 - 25 | 2010 | 63 | ||
| GenBank mRNA | AK101127.1 | - | 25 - 26 | 2113 | 63 | |
More Information
Set Membership: |
|

LOC4339215
Genomic Map