Assay Details
| Gene Symbol: LOC4339345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783937.2 | XP_015639423.1 | 12 - 13 | 1143 | 78 | |
| GenBank mRNA | AK067224.1 | - | 12 - 13 | 1102 | 78 | |
| AK070919.1 | - | 12 - 13 | 1102 | 78 | ||
| AK101078.1 | - | 7 - 8 | 640 | 78 | ||
More Information
Set Membership: |
|

LOC4339345
Genomic Map