Assay Details
| Gene Symbol: LOC4339345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783937.2 | XP_015639423.1 | 18 - 19 | 2060 | 135 | |
| GenBank mRNA | AK067224.1 | - | 18 - 19 | 2019 | 135 | |
| AK101078.1 | - | 13 - 14 | 1555 | 135 | ||
More Information
Set Membership: |
|

LOC4339345
Genomic Map