Assay Details
| Gene Symbol: LOC4339345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783937.2 | XP_015639423.1 | 19 - 20 | 2155 | 103 | |
| GenBank mRNA | AK067224.1 | - | 19 - 20 | 2114 | 103 | |
| AK101078.1 | - | 14 - 15 | 1650 | 103 | ||
More Information
Set Membership: |
|

LOC4339345
Genomic Map