Assay Details
| Gene Symbol: LOC4339345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783937.2 | XP_015639423.1 | 21 - 22 | 2324 | 107 | |
| GenBank mRNA | AK067224.1 | - | 21 - 22 | 2283 | 107 | |
| AK101078.1 | - | 15 - 16 | 1906 | 107 | ||
More Information
Set Membership: |
|

LOC4339345
Genomic Map