Assay Details
| Gene Symbol: LOC4339345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783937.2 | XP_015639423.1 | 2 - 3 | 305 | 87 | |
| GenBank mRNA | AK067224.1 | - | 2 - 3 | 264 | 87 | |
| AK070919.1 | - | 2 - 3 | 264 | 87 | ||
More Information
Set Membership: |
|

LOC4339345
Genomic Map