Assay Details
| Gene Symbol: LOC4339353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783988.2 | XP_015639474.1 | 9 - 10 | 2332 | 76 | |
| Gene Symbol: LOC107278650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK065361.1 | - | 15 - 16 | 1603 | 76 | |
More Information
Set Membership: |
|

LOC4339353
Genomic Map