Assay Details
| Gene Symbol: LOC4339487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784667.2 | XP_015640153.1 | 2 | 1146 | 66 | |
| GenBank mRNA | AK242655.1 | - | 2 | 1135 | 66 | |
More Information
Set Membership: |
|

LOC4339487
Genomic Map