Assay Details
| Gene Symbol: LOC4339491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781940.2 | XP_015637426.1 | 1 - 2 | 325 | 79 | |
| GenBank mRNA | AK071113.1 | - | 1 - 2 | 113 | 79 | |
| AK103912.1 | - | 1 - 2 | 69 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015781940.2 transcripts.
Set Membership: |
|

LOC4339491
Genomic Map