Assay Details
| Gene Symbol: LOC4339491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781940.2 | XP_015637426.1 | 4 - 5 | 583 | 95 | |
| GenBank mRNA | AK071113.1 | - | 4 - 5 | 371 | 95 | |
| AK103912.1 | - | 4 - 5 | 327 | 95 | ||
More Information
Set Membership: |
|

LOC4339491
Genomic Map