Assay Details
| Gene Symbol: LOC4339491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781940.2 | XP_015637426.1 | 7 - 8 | 1039 | 70 | |
| GenBank mRNA | AK071113.1 | - | 7 - 8 | 827 | 70 | |
| AK103912.1 | - | 7 - 8 | 783 | 70 | ||
More Information
Set Membership: |
|

LOC4339491
Genomic Map