Assay Details
| Gene Symbol: LOC4339541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784993.2 | XP_015640479.1 | 6 - 7 | 765 | 101 | |
| XM_015784994.2 | XP_015640480.1 | 6 - 7 | 764 | 101 | ||
| GenBank mRNA | AK105695.1 | - | 6 - 7 | 659 | 101 | |
| AK121660.1 | - | 6 - 7 | 735 | 101 | ||
More Information
Set Membership: |
|

LOC4339541
Genomic Map