Assay Details
| Gene Symbol: LOC4339718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784986.1 | XP_015640472.1 | 13 - 14 | 1699 | 101 | |
| XM_015784987.1 | XP_015640473.1 | 14 - 15 | 1947 | 101 | ||
| GenBank mRNA | AK100910.1 | - | 13 - 14 | 1706 | 101 | |
| EU267957.1 | - | 13 - 14 | 1415 | 101 | ||
More Information
Set Membership: |
|

LOC4339718
Genomic Map