Assay Details
| Gene Symbol: LOC4339718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784986.1 | XP_015640472.1 | 14 - 15 | 1808 | 74 | |
| XM_015784987.1 | XP_015640473.1 | 15 - 16 | 2056 | 74 | ||
| GenBank mRNA | AK100910.1 | - | 14 - 15 | 1815 | 74 | |
| D50317.1 | - | 14 - 15 | 1534 | 74 | ||
| EU267957.1 | - | 14 - 15 | 1524 | 74 | ||
More Information
Set Membership: |
|

LOC4339718
Genomic Map