Assay Details
| Gene Symbol: LOC4339718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784986.1 | XP_015640472.1 | 3 - 4 | 736 | 83 | |
| XM_015784987.1 | XP_015640473.1 | 4 - 5 | 984 | 83 | ||
| GenBank mRNA | AK100910.1 | - | 3 - 4 | 743 | 83 | |
| D50317.1 | - | 3 - 4 | 462 | 83 | ||
| EU267957.1 | - | 3 - 4 | 452 | 83 | ||
More Information
Set Membership: |
|

LOC4339718
Genomic Map