Assay Details
| Gene Symbol: LOC4339718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784986.1 | XP_015640472.1 | 6 - 7 | 1085 | 114 | |
| XM_015784987.1 | XP_015640473.1 | 7 - 8 | 1333 | 114 | ||
| GenBank mRNA | AK100910.1 | - | 6 - 7 | 1092 | 114 | |
| D50317.1 | - | 6 - 7 | 811 | 114 | ||
| EU267957.1 | - | 6 - 7 | 801 | 114 | ||
More Information
Set Membership: |
|

LOC4339718
Genomic Map