Assay Details
| Gene Symbol: LOC4339718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784986.1 | XP_015640472.1 | 7 - 8 | 1146 | 110 | |
| XM_015784987.1 | XP_015640473.1 | 8 - 9 | 1394 | 110 | ||
| GenBank mRNA | AK100910.1 | - | 7 - 8 | 1153 | 110 | |
| D50317.1 | - | 7 - 8 | 872 | 110 | ||
| EU267957.1 | - | 7 - 8 | 862 | 110 | ||
More Information
Set Membership: |
|

LOC4339718
Genomic Map