Assay Details
| Gene Symbol: LOC4340043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788881.2 | XP_015644367.1 | 13 - 14 | 1712 | 64 | |
| XM_015788882.2 | XP_015644368.1 | 13 - 14 | 1712 | 64 | ||
| XM_015788883.2 | XP_015644369.1 | 13 - 14 | 1706 | 64 | ||
| XM_015788884.2 | XP_015644370.1 | 13 - 14 | 1712 | 64 | ||
| XM_015788885.2 | XP_015644371.1 | 13 - 14 | 1597 | 64 | ||
| XM_015788886.2 | XP_015644372.1 | 14 - 15 | 1509 | 64 | ||
| GenBank mRNA | AK065081.1 | - | 13 - 14 | 1691 | 64 | |
More Information
Set Membership: |
|

LOC4340043
Genomic Map