Assay Details
| Gene Symbol: LOC4340043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788881.2 | XP_015644367.1 | 17 - 18 | 2133 | 149 | |
| XM_015788882.2 | XP_015644368.1 | 17 - 18 | 2133 | 149 | ||
| XM_015788883.2 | XP_015644369.1 | 17 - 18 | 2127 | 149 | ||
| XM_015788884.2 | XP_015644370.1 | 17 - 18 | 2133 | 149 | ||
| XM_015788885.2 | XP_015644371.1 | 17 - 18 | 2018 | 149 | ||
| XM_015788886.2 | XP_015644372.1 | 18 - 19 | 1930 | 149 | ||
| GenBank mRNA | AK065081.1 | - | 17 - 18 | 2112 | 149 | |
More Information
Set Membership: |
|

LOC4340043
Genomic Map