Assay Details
| Gene Symbol: LOC4340043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788881.2 | XP_015644367.1 | 1 - 2 | 110 | 94 | |
| XM_015788882.2 | XP_015644368.1 | 1 - 2 | 110 | 94 | ||
| XM_015788883.2 | XP_015644369.1 | 1 - 2 | 110 | 94 | ||
| XM_015788884.2 | XP_015644370.1 | 1 - 2 | 110 | 94 | ||
| GenBank mRNA | AK065081.1 | - | 1 - 2 | 89 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788881.2, XM_015788882.2, XM_015788883.2, XM_015788884.2 transcripts.
Set Membership: |
|

LOC4340043
Genomic Map