Assay Details
| Gene Symbol: LOC4340422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788981.2 | XP_015644467.1 | 2 - 3 | 742 | 53 | |
| GenBank mRNA | AK109607.1 | - | 2 - 3 | 626 | 53 | |
More Information
Set Membership: |
|

LOC4340422
Genomic Map