Assay Details
| Gene Symbol: LOC4340506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785626.2 | XP_015641112.1 | 8 - 9 | 1082 | 79 | |
| XM_015785627.2 | XP_015641113.1 | 8 - 9 | 1066 | 79 | ||
| XM_015785628.2 | XP_015641114.1 | 8 - 9 | 824 | 79 | ||
| XM_015785629.2 | XP_015641115.1 | 8 - 9 | 1082 | 79 | ||
More Information
Set Membership: |
|

LOC4340506
Genomic Map