Assay Details
| Gene Symbol: LOC4340523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788766.2 | XP_015644252.1 | 1 | 721 | 65 | |
| GenBank mRNA | AK108233.1 | - | 1 | 652 | 65 | |
More Information
Set Membership: |
|

LOC4340523
Genomic Map