Assay Details
| Gene Symbol: LOC9267065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786969.2 | XP_015642455.1 | 1 - 2 | 146 | 68 | |
| XM_015786972.2 | XP_015642458.1 | 1 - 2 | 249 | 68 | ||
| GenBank mRNA | AK122137.1 | - | 1 - 2 | 251 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015786969.2, XM_015786972.2 transcripts.
Set Membership: |
|

LOC9267065
Genomic Map