Assay Details
| Gene Symbol: LOC9267065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786972.2 | XP_015642458.1 | 4 - 5 | 634 | 74 | |
| GenBank mRNA | AK106539.1 | - | 1 - 2 | 90 | 74 | |
| AK122137.1 | - | 4 - 5 | 636 | 74 | ||
More Information
Set Membership: |
|

LOC9267065
Genomic Map