Assay Details
| Gene Symbol: LOC9267065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786968.2 | XP_015642454.1 | 38 - 39 | 13285 | 58 | |
| XM_015786969.2 | XP_015642455.1 | 38 - 39 | 13211 | 58 | ||
| XM_015786970.2 | XP_015642456.1 | 38 - 39 | 13085 | 58 | ||
| XM_015786972.2 | XP_015642458.1 | 39 - 40 | 13227 | 58 | ||
| GenBank mRNA | AK060815.1 | - | 1 - 2 | 1007 | 58 | |
| AK066536.1 | - | 3 - 4 | 2466 | 58 | ||
More Information
Set Membership: |
|

LOC9267065
Genomic Map