Assay Details
| Gene Symbol: LOC4341499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788538.2 | XP_015644024.1 | 4 - 5 | 539 | 99 | |
| XM_015788539.2 | XP_015644025.1 | 5 - 6 | 646 | 99 | ||
| GenBank mRNA | AK071731.1 | - | 4 - 5 | 499 | 99 | |
More Information
Set Membership: |
|

LOC4341499
Genomic Map