Assay Details
| Gene Symbol: LOC4341499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788538.2 | XP_015644024.1 | 5 - 6 | 592 | 65 | |
| XM_015788539.2 | XP_015644025.1 | 6 - 7 | 699 | 65 | ||
| GenBank mRNA | AK071731.1 | - | 5 - 6 | 552 | 65 | |
More Information
Set Membership: |
|

LOC4341499
Genomic Map