Assay Details
| Gene Symbol: LOC4341544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785538.2 | XP_015641024.1 | 4 - 5 | 572 | 83 | |
| XM_015785539.2 | XP_015641025.1 | 4 - 5 | 590 | 83 | ||
| GenBank mRNA | AK059836.1 | - | 4 - 5 | 519 | 83 | |
| AK071563.1 | - | 4 - 5 | 506 | 83 | ||
More Information
Set Membership: |
|

LOC4341544
Genomic Map