Assay Details
| Gene Symbol: LOC4341767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787384.2 | XP_015642870.1 | 2 - 3 | 582 | 72 | |
| GenBank mRNA | AK071785.1 | - | 2 - 3 | 554 | 72 | |
| AK111631.1 | - | 2 - 3 | 551 | 72 | ||
| AK120152.1 | - | 1 - 2 | 252 | 72 | ||
More Information
Set Membership: |
|

LOC4341767
Genomic Map