Assay Details
| Gene Symbol: LOC4341767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787384.2 | XP_015642870.1 | 3 - 4 | 1222 | 60 | |
| XM_015787385.2 | XP_015642871.1 | 2 - 3 | 1174 | 60 | ||
| GenBank mRNA | AK071785.1 | - | 3 - 4 | 1194 | 60 | |
| AK111631.1 | - | 3 - 4 | 1191 | 60 | ||
| AK120152.1 | - | 2 - 3 | 892 | 60 | ||
More Information
Set Membership: |
|

LOC4341767
Genomic Map