Assay Details
| Gene Symbol: LOC4341767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787384.2 | XP_015642870.1 | 7 - 8 | 1845 | 99 | |
| XM_015787385.2 | XP_015642871.1 | 6 - 7 | 1797 | 99 | ||
| GenBank mRNA | AK111631.1 | - | 7 - 8 | 1814 | 99 | |
| AK120152.1 | - | 6 - 7 | 1515 | 99 | ||
More Information
Set Membership: |
|

LOC4341767
Genomic Map