Assay Details
| Gene Symbol: LOC4341897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788118.2 | XP_015643604.1 | 4 - 5 | 480 | 134 | |
| XM_015788119.2 | XP_015643605.1 | 4 - 5 | 427 | 134 | ||
| XM_015788121.2 | XP_015643607.1 | 5 - 6 | 474 | 134 | ||
| GenBank mRNA | AK063450.1 | - | 4 - 5 | 427 | 134 | |
| AK101144.1 | - | 4 - 5 | 421 | 134 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788121.2 transcripts.
Set Membership: |
|

LOC4341897
Genomic Map