Assay Details
| Gene Symbol: LOC4341897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788118.2 | XP_015643604.1 | 2 - 3 | 310 | 138 | |
| XM_015788119.2 | XP_015643605.1 | 2 - 3 | 257 | 138 | ||
| GenBank mRNA | AK063450.1 | - | 2 - 3 | 257 | 138 | |
| AK101144.1 | - | 2 - 3 | 251 | 138 | ||
More Information
Set Membership: |
|

LOC4341897
Genomic Map