Assay Details
| Gene Symbol: LOC4341897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788118.2 | XP_015643604.1 | 9 - 10 | 1131 | 81 | |
| XM_015788119.2 | XP_015643605.1 | 9 - 10 | 1078 | 81 | ||
| XM_015788121.2 | XP_015643607.1 | 10 - 11 | 1125 | 81 | ||
| GenBank mRNA | AK063450.1 | - | 9 - 10 | 1078 | 81 | |
| AK101144.1 | - | 9 - 10 | 1072 | 81 | ||
More Information
Set Membership: |
|

LOC4341897
Genomic Map