Assay Details
| Gene Symbol: LOC4342609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791740.2 | XP_015647226.1 | 1 - 2 | 805 | 76 | |
| GenBank mRNA | AK063957.1 | - | 1 - 2 | 685 | 76 | |
| AK073755.1 | - | 1 - 2 | 806 | 76 | ||
| AK107609.1 | - | 1 - 2 | 802 | 76 | ||
More Information
Set Membership: |
|

LOC4342609
Genomic Map