Assay Details
| Gene Symbol: LOC4342609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791740.2 | XP_015647226.1 | 2 - 3 | 1029 | 78 | |
| GenBank mRNA | AK073755.1 | - | 2 - 3 | 1030 | 78 | |
| AK107609.1 | - | 2 - 3 | 1026 | 78 | ||
More Information
Set Membership: |
|

LOC4342609
Genomic Map