Assay Details
| Gene Symbol: LOC4342702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791396.1 | XP_015646882.1 | 5 - 6 | 758 | 63 | |
| GenBank mRNA | AK059111.1 | - | 5 - 6 | 765 | 63 | |
| AK099319.1 | - | 5 - 6 | 758 | 63 | ||
More Information
Set Membership: |
|

LOC4342702
Genomic Map